Merck
  • Exosomal miR-673-5p from fibroblasts promotes Schwann cell-mediated peripheral neuron myelination by targeting the TSC2/mTORC1/SREBP2 axis.
Exosomal miR-673-5p from fibroblasts promotes Schwann cell-mediated peripheral neuron myelination by targeting the TSC2/mTORC1/SREBP2 axis.
The Journal of biological chemistry (2022-02-14)
Yahong Zhao, Yunyun Liang, Zhixin Xu, Jina Liu, Xiaoyu Liu, Jinyu Ma, Cheng Sun, Yumin Yang
ABSTRACT
Peripheral myelination is a complicated process, wherein Schwann cells (SCs) promote the formation of the myelin sheath around the axons of peripheral neurons. Fibroblasts are the second resident cells in the peripheral nerves; however, the precise function of fibroblasts in SC-mediated myelination has rarely been examined. Here, we show that exosomes derived from fibroblasts boost myelination-related gene expression in SCs. We used exosome sequencing, together with bioinformatic analysis, to demonstrate that exosomal microRNA miR-673-5p is capable of stimulating myelin gene expression in SCs. Subsequent functional studies revealed that miR-673-5p targets the regulator of mechanistic target of the rapamycin (mTOR) complex 1 (mTORC1) tuberous sclerosis complex 2 in SCs, leading to the activation of downstream signaling pathways including mTORC1 and sterol-regulatory element binding protein 2. In vivo experiments further confirmed that miR-673-5p activates the tuberous sclerosis complex 2/mTORC1/sterol-regulatory element binding protein 2 axis, thus promoting the synthesis of cholesterol and related lipids and subsequently accelerating myelin sheath maturation in peripheral nerves. Overall, our findings revealed exosome-mediated cross talk between fibroblasts and SCs that plays a pivotal role in peripheral myelination. We propose that exosomes derived from fibroblasts and miR-673-5p might be useful for promoting peripheral myelination in translational medicine.
Sigma-Aldrich
Cholesterol Extraction Kit, sufficient for 40 extractions
Sigma-Aldrich
Cholesterol Quantitation Kit, sufficient for 100 colorimetric or fluorometric tests