Merck
  • Single-cell transcriptomics links malignant T cells to the tumor immune landscape in cutaneous T cell lymphoma.
Single-cell transcriptomics links malignant T cells to the tumor immune landscape in cutaneous T cell lymphoma.
Nature communications (2022-03-05)
Xiangjun Liu, Shanzhao Jin, Simeng Hu, Ruoyan Li, Haihao Pan, Yi Liu, Pan Lai, Deshu Xu, Jingru Sun, Ziyang Liu, Yumei Gao, Yifan Zhao, Fengjie Liu, Yu Xiao, Yingyi Li, Yujie Wen, Zhuojing Chen, Bufang Xu, Yuchieh Lin, Menglong Ran, Qianxi Li, Shuxia Yang, Hang Li, Ping Tu, Muzlifah Haniffa, Sarah A Teichmann, Fan Bai, Yang Wang
ABSTRACT
Cutaneous T cell lymphoma (CTCL) represents a heterogeneous group of non-Hodgkin lymphoma distinguished by the presence of clonal malignant T cells. The heterogeneity of malignant T cells and the complex tumor microenvironment remain poorly characterized. With single-cell RNA analysis and bulk whole-exome sequencing on 19 skin lesions from 15 CTCL patients, we decipher the intra-tumor and inter-lesion diversity of CTCL patients and propose a multi-step tumor evolution model. We further establish a subtyping scheme based on the molecular features of malignant T cells and their pro-tumorigenic microenvironments: the TCyEM group, demonstrating a cytotoxic effector memory T cell phenotype, shows more M2 macrophages infiltration, while the TCM group, featured by a central memory T cell phenotype and adverse patient outcome, is infiltrated by highly exhausted CD8+ reactive T cells, B cells and Tregs with suppressive activities. Our results establish a solid basis for understanding the nature of CTCL and pave the way for future precision medicine for CTCL patients.
Sigma-Aldrich
Anti-CD45RA Antibody, Clone MEM 56, clone MEM 56, from mouse