Merck
  • Analgesic and Antipyretic Activities of Ethyl Acetate Fraction Tablet of Andrographis paniculata in Animal Models.
Analgesic and Antipyretic Activities of Ethyl Acetate Fraction Tablet of Andrographis paniculata in Animal Models.
Evidence-based complementary and alternative medicine : eCAM (2021-03-23)
Hilkatul Ilmi, Irfan Rayi Pamungkas, Lidya Tumewu, Achmad Fuad Hafid, Aty Widyawaruyanti
ABSTRACT
To determine the analgesic and antipyretic activities of a tablet derived from Andrographis paniculata ethyl acetate fraction (AS201-01) in animal models. The tablet derived from AS201-01 contains an equivalent of 35 mg andrographolide per tablet. Analgesic activity was determined using an acetic acid-induced writhing test on adult male mice. A writhe was recorded by a stopwatch and was defined as the stretching of the abdomen and/or stretching of at least one hind limb. For the determination of antipyretic activity, pyrexia was induced by subcutaneous injection of 15% w/v Brewer's yeast into adult male rats. Rectal temperature was monitored at 1, 2, 3, and 4 hours after treatment. The results showed that the AS201-01 tablet had analgesic and antipyretic activity. In the acetic acid-induced writhing model, AS201-01 tablet exhibited significant analgesic effect with a 66.73% reduction in writhing response at a dose of 50 mg andrographolide/kg body weight compared to the negative control group. The tablet also showed a significant antipyretic effect. The maximum antipyretic effect was observed after the third hour of administration of the AS201-01 tablet at a dose of 100 mg andrographolide/kg body weight. Tablet of Andrographis paniculata ethyl acetate fraction (AS201-01) exhibited analgesic and antipyretic activities.
Sigma-Aldrich
Yeast, powder, slightly brown