- Cryopreservation of mouse spermatozoa and in vitro fertilization.
Cryopreservation of mouse spermatozoa and in vitro fertilization.
Methods in molecular biology (Clifton, N.J.) (2010-11-17)
Naomi Nakagata
PMID21080274
ABSTRACT
Cryopreservation of mouse spermatozoa has become the foremost technique for preserving large numbers of different strains of mice with induced mutations. Recently, we have established procedures for cryopreservation of mouse spermatozoa and in vitro fertilization using cryopreserved spermatozoa to obtain a relatively high fertilization rate. This chapter attempts to show these procedures in simple terms.
Sigma-Aldrich
Phenol red solution, 0.5%, liquid, sterile-filtered, BioReagent, suitable for cell culture