Merck
  • Determination and uptake of abamectin and difenoconazole in the stingless bee Melipona scutellaris Latreille, 1811 via oral and topic acute exposure.
Determination and uptake of abamectin and difenoconazole in the stingless bee Melipona scutellaris Latreille, 1811 via oral and topic acute exposure.
Environmental pollution (Barking, Essex : 1987) (2020-08-19)
Fernanda Scavassa Ribeiro do Prado, Dayana Moscardi Dos Santos, Thiessa Maramaldo de Almeida Oliveira, José Augusto Micheletti Burgarelli, Janete Brigante Castele, Eny Maria Vieira
ABSTRACT
Bees are considered as important providers of ecosystem services, acting via pollination process in crops and native plants, and contributing significantly to the maintenance of biodiversity. However, the decrease of bee's population has been observed worldwide and besides other factors, this collapse is also related to the extensive use of pesticides. In this sense, studies involving the assessment of adverse effects and the uptake of pesticides by bees are of great concern. This work presents an analytical method for the determination of the insecticide abamectin and the fungicide difenoconazole in the stingless bee Melipona scutellaris exposed via oral and topic to endpoints concentrations of active ingredients (a.i.) alone and in commercial formulations and the discussion about its mortality and uptake. For this purpose, QuEChERS (Quick, Easy, Cheap, Efficient, Rugged and Safe) acetate modified method was used for extraction and pesticides were determined by LC-MS/MS. The validation parameters have included: a linear range between 0.01 and 1.00 μg mL-1; and LOD and LOQ of 0.038 and 0.076 μg g-1 for abamectin and difenoconazole, respectively. The uptake of tested pesticides via oral and topic was verified by the accumulation in adult forager bees, mainly when the commercial product was tested. Mortality was observed to be higher in oral exposure than in topic tests for both pesticides. For abamectin in a commercial formulation (a.i.) no differences were observed for oral or topic exposure. On the other hand, for difenoconazole, topic exposure had demonstrated higher accumulation in bees, according to the increase of received dose. Through the results, uptake and the possible consequences of bioaccumulated pesticides are also discussed and can contribute to the knowledge about the risks involving the exposure of bees to these compounds.
Product Description
Supelco
Difenoconazol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.