Merck
  • Murine UDP-GlcNAc:lysosomal enzyme N-acetylglucosamine-1-phosphotransferase lacking the gamma-subunit retains substantial activity toward acid hydrolases.
Murine UDP-GlcNAc:lysosomal enzyme N-acetylglucosamine-1-phosphotransferase lacking the gamma-subunit retains substantial activity toward acid hydrolases.
The Journal of biological chemistry (2007-07-27)
Wang-Sik Lee, Bobby Joe Payne, Claire M Gelfman, Peter Vogel, Stuart Kornfeld
ABSTRACT
UDP-GlcNAc:lysosomal enzyme N-acetylglucosamine-1-phosphotransferase (GlcNAc-1-phosphotransferase) mediates the first step in the synthesis of the mannose 6-phosphate recognition marker on acid hydrolases. The transferase exists as an alpha(2)beta(2)gamma(2) hexameric complex with the alpha- and beta-subunits derived from a single precursor molecule. The catalytic function of the transferase is attributed to the alpha- and beta-subunits, whereas the gamma-subunit is believed to be involved in the recognition of a conformation-dependent protein determinant common to acid hydrolases. Using knock-out mice with mutations in either the alpha/beta gene or the gamma gene, we show that disruption of the alpha/beta gene completely abolishes phosphorylation of high mannose oligosaccharides on acid hydrolases whereas knock-out of the gamma gene results in only a partial loss of phosphorylation. These findings demonstrate that the alpha/beta-subunits, in addition to their catalytic function, have some ability to recognize acid hydrolases as specific substrates. This process is enhanced by the gamma-subunit.
Sigma-Aldrich
4-Methylumbelliferyl α-D-mannopyranoside, ≥97% (HPLC)