Merck
  • Saffloflavone, a new flavonoid from the flowers of Carthamus tinctorius L. and its cardioprotective activity.
Saffloflavone, a new flavonoid from the flowers of Carthamus tinctorius L. and its cardioprotective activity.
Natural product research (2021-01-13)
Meng-Na Wang, Yan-Gang Cao, Ya-Xin Wei, Ying-Jie Ren, Yan-Ling Liu, Xu Chen, Chen He, Xiao-Ke Zheng, Wei-Sheng Feng
ABSTRACT
A new flavonoid, saffloflavone , along with six known compounds, kaempferol-3-O-rutinoside, kaempferol-3-O-sophoroside, quercetin-3-O-β-d-glucoside, quercetin-7-O-β-d-glucoside, luteolin-7-O-β-d-glucoside and kaempferol 3-O-β-d-glucoside were isolated from the flowers of Carthamus tinctorius L. All the structures were determined by interpretation of their spectroscopic data. The cardioprotective effects of all the isolates against oxidative stress of H9c2 cells induced by H2O2 were investigated. The results showed that compounds 4-6 exhibited protective effects against of H9c2 cells injury induced by H2O2.
Sigma-Aldrich
Luteolin 7-O-β-D-glucoside, ≥98.0% (HPLC)
Pricing and availability is not currently available.