Merck
D-19466, a new cyclobutane-platinum complex with antitumor activity.
Journal of cancer research and clinical oncology (1990-01-01)
R Voegeli, W Schumacher, J Engel, J Respondek, P Hilgard
ABSTRACT
D-19466, a new platinum complex, was characterized. It showed no nephrotoxic side-effects as determined by the measurement of blood urea. It was cytotoxic in vitro for tumor cells in concentrations comparable to or lower than cytotoxic concentrations of cisplatin. It had excellent anticancer activity in vivo against a number of murine experimental tumors, including a cisplatin-resistant P388 line. Clinical evaluation of this compound has therefore been initiated.
Sigma-Aldrich
Lobaplatin, ≥98% (NMR)