Merck
  • Dysregulated healing responses in diabetic wounds occur in the early stages postinjury.
Dysregulated healing responses in diabetic wounds occur in the early stages postinjury.
Journal of molecular endocrinology (2020-12-23)
Kiara Boodhoo, Mare Vlok, David L Tabb, Kathryn H Myburgh, Mari van de Vyver
ABSTRACT
Chronic wounds are a serious and debilitating complication of diabetes. A better understanding of the dysregulated healing responses following injury will provide insight into the optimal time frame for therapeutic intervention. In this study, a direct comparison was done between the healing dynamics and the proteome of acute and obese diabetic wounds on days 2 and 7 following injury. Full thickness excisional wounds were induced on obese diabetic (B6.Cg-lepob/J, ob/ob, n = 14) (blood glucose 423.25 ± 127.92 mg/dL) and WT control (C57BL/6J, n = 14) (blood glucose 186.67 ± 24.5 mg/dL) mice. Histological analysis showed no signs of healing in obese DM wounds whereas complete wound closure/re-epithelisation, the formation of granulation tissue and signs of re-vascularisation, was evident in acute wounds on day 7. In obese DM wounds, substance P deficiency and increased MMP-9 activity on day 2 coincided with increased cytokine/chemokine levels within wound fluid. LC-MS/MS identified 906 proteins, of which 23 (Actn3, Itga6, Epb41, Sncg, Nefm, Rsp18, Rsp19, Rpl22, Macroh2a1, Rpn1, Ppib, Snrnp70, Ddx5, Eif3g, Tpt1, FABP5, Cavin1, Stfa1, Stfa3, Cycs, Tkt, Mb, Chmp2a) were differentially expressed in wounded tissue on day 2 (P < 0.05; more than two-fold) and 6 (Cfd, Ptms, Hp, Hmga1, Cbx3, Syap1) (P < 0.05; more than two-fold) on day 7. A large number of dysregulated proteins on day 2 was associated with an inability to progress into the proliferative stage of healing and suggest that early intervention might be pivotal for effective healing outcomes. The proteomic approach highlighted the complexity of obese DM wounds in which the dysregulation involves multiple regulatory pathways and biological processes.
Sigma-Aldrich
Neprilysin/CD10 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)