Merck
  • Immobilized Enzymes on Gold Nanoparticles: From Enhanced Stability to Cleaning of Heritage Textiles.
Immobilized Enzymes on Gold Nanoparticles: From Enhanced Stability to Cleaning of Heritage Textiles.
ACS applied bio materials (2019-11-18)
Francesca Gherardi, Lyudmila Turyanska, Enrico Ferrari, Nicola Weston, Michael W Fay, Belinda J Colston
ABSTRACT
Enzyme-based treatments are used in heritage conservation for the effective removal of glues and other damaging organic layers from the surfaces of historic and artistic works. Despite their potential, however, the application of enzymatic treatments is currently limited because of their poor efficiency and low operational and environmental stability. We demonstrate the use of α-amylase immobilized on gold nanoparticles to improve the efficacy of enzymatic treatments enhancing both the reactivity and the stability of the formulations. Gold nanoparticles coated with α-amylase exhibit significant advantages compared to free enzymes. We report up to 5× greater resistance to environmental changes, up to 2× higher efficacy toward removal of starch-based glues from textiles and deeper penetration through the fibers, without causing damage or inducing salt precipitation. These results offer exciting prospects for the development of enzymatic formulations, both for heritage conservation and the wider application of enzymes, such as in medicine, the detergent industry, and green chemistry.
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Pricing and availability is not currently available.