Merck
  • CDH6-activated αIIbβ3 crosstalks with α2β1 to trigger cellular adhesion and invasion in metastatic ovarian and renal cancers.
CDH6-activated αIIbβ3 crosstalks with α2β1 to trigger cellular adhesion and invasion in metastatic ovarian and renal cancers.
Molecular oncology (2021-03-15)
Rubén A Bartolomé, Javier Robles, Ángela Martin-Regalado, Laura Pintado-Berninches, Miranda Burdiel, Marta Jaén, Carmen Aizpurúa, Juan I Imbaud, José Ignacio Casal
ABSTRACT
Cadherin 6 (CDH6) is significantly overexpressed in advanced ovarian and renal cancers. However, the role of CDH6 in cancer metastasis is largely unclear. Here, we investigated the impact of CDH6 expression on integrin-mediated metastatic progression. CDH6 preferentially bound to αIIbβ3 integrin, a platelet receptor scarcely expressed in cancer cells, and this interaction was mediated through the cadherin Arginine-glycine-aspartic acid (RGD) motif. Furthermore, CDH6 and CDH17 were found to interact with α2β1 in αIIbβ3low cells. Transient silencing of CDH6, ITGA2B, or ITGB3 genes caused a significant loss of proliferation, adhesion, invasion, and lung colonization through the downregulation of SRC, FAK, AKT, and ERK signaling. In ovarian and renal cancer cells, integrin αIIbβ3 activation appears to be a prerequisite for proper α2β1 activation. Interaction of αIIbβ3 with CDH6, and subsequent αIIbβ3 activation, promoted activation of α2β1 and cell adhesion in ovarian and renal cancer cells. Additionally, monoclonal antibodies specific to the cadherin RGD motif and clinically approved αIIbβ3 inhibitors could block pro-metastatic activity in ovarian and renal tumors. In summary, the interaction between CDH6 and αIIbβ3 regulates α2β1-mediated adhesion and invasion of ovarian and renal cancer metastatic cells and constitutes a therapeutic target of broad potential for treating metastatic progression.
Sigma-Aldrich
Anti-LIBS2 epitope (Integrin beta3 subunit) Antibody, clone ab62, clone ab62, from mouse