Merck
  • Infrared spectroscopy of the alanine dipeptide analog in liquid water with DFT-MD. Direct evidence for P(II)/beta conformations.
Infrared spectroscopy of the alanine dipeptide analog in liquid water with DFT-MD. Direct evidence for P(II)/beta conformations.
Physical chemistry chemical physics : PCCP (2010-06-12)
Marie-Pierre Gaigeot
ABSTRACT
Following our previous work [J. Phys. Chem. B. Lett., 2009, 113, 10059], DFT-based molecular dynamics (DFTMD) simulations of 2-Ala peptide (i.e. Ac-Ala-NHMe dialanine peptide analog with methyl group caps at the extremities) immersed in liquid water at room temperature are reported. Our goal here is the theoretical calculation of the infrared spectrum of aqueous 2-Ala, in order to provide a definitive understanding of the average conformation adopted by this peptide in the liquid phase, taking into account solute and solvent at the same theoretical level of representation. We find that the experimental Amide I-II band predominantly results from a mixture of partially unfolded P(II) and unfolded beta conformational equilibrium of aqueous 2-Ala at room temperature.
Sigma-Aldrich
Ala-Gly
Sigma-Aldrich
D-Ala-D-Ala
Sigma-Aldrich
Ala-Leu
Sigma-Aldrich
Ala-Phe
Sigma-Aldrich
Ala-Pro hydrate
Sigma-Aldrich
Ala-Ala
Sigma-Aldrich
Ala-Tyr