Merck
  • Live tumor imaging shows macrophage induction and TMEM-mediated enrichment of cancer stem cells during metastatic dissemination.
Live tumor imaging shows macrophage induction and TMEM-mediated enrichment of cancer stem cells during metastatic dissemination.
Nature communications (2021-12-17)
Ved P Sharma, Binwu Tang, Yarong Wang, Camille L Duran, George S Karagiannis, Emily A Xue, David Entenberg, Lucia Borriello, Anouchka Coste, Robert J Eddy, Gina Kim, Xianjun Ye, Joan G Jones, Eli Grunblatt, Nathan Agi, Sweta Roy, Gargi Bandyopadhyaya, Esther Adler, Chinmay R Surve, Dominic Esposito, Sumanta Goswami, Jeffrey E Segall, Wenjun Guo, John S Condeelis, Lalage M Wakefield, Maja H Oktay
ABSTRACT
Cancer stem cells (CSCs) play an important role during metastasis, but the dynamic behavior and induction mechanisms of CSCs are not well understood. Here, we employ high-resolution intravital microscopy using a CSC biosensor to directly observe CSCs in live mice with mammary tumors. CSCs display the slow-migratory, invadopod-rich phenotype that is the hallmark of disseminating tumor cells. CSCs are enriched near macrophages, particularly near macrophage-containing intravasation sites called Tumor Microenvironment of Metastasis (TMEM) doorways. Substantial enrichment of CSCs occurs on association with TMEM doorways, contributing to the finding that CSCs represent >60% of circulating tumor cells. Mechanistically, stemness is induced in non-stem cancer cells upon their direct contact with macrophages via Notch-Jagged signaling. In breast cancers from patients, the density of TMEM doorways correlates with the proportion of cancer cells expressing stem cell markers, indicating that in human breast cancer TMEM doorways are not only cancer cell intravasation portals but also CSC programming sites.
Sigma-Aldrich
Anti-HES-1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
AldeRed® ALDH Detection Assay