Merck
  • Quantitation of fatty acid ethyl esters in human meconium by an improved liquid chromatography/tandem mass spectrometry.
Quantitation of fatty acid ethyl esters in human meconium by an improved liquid chromatography/tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-07-08)
Ho-Seok Kwak, Young-Sun Kang, Ki-Ok Han, Jong-Taek Moon, Young-Chul Chung, June-Seek Choi, Jung-Yeol Han, Moon-Young Kim, E Yadira Velázquez-Armenta, Alejandro A Nava-Ocampo
ABSTRACT
This paper reports the development and validation of an improved assay for quantitation of fatty acid ethyl esters (FAEEs) in human meconium using liquid chromatography/tandem mass spectrometry (LC-MS/MS). FAAEs (ethyl laurate, ethyl myristate, ethyl palmitate, ethyl palmitoleate, ethyl stearate, ethyl oleate, ethyl linoleate, ethyl linolenate, and ethyl arachidonate) and the internal standard (I.S.), ethyl heptadecanoate, were separated by reverse phase HPLC and quantified by MS/MS using electrospray ionization (ESI) and multiple reaction monitoring (MRM) in the positive ionization mode. The absolute recovery of FAEEs varied from 55+/-10% for 0.33 nmol/g (100 ng/g) of ethyl linoleate up to 86+/-8% for 1.55 nmol/g (500 ng/g) of ethyl miristate. The LODs and LOQs varied from 0.01 to 0.08 nmol/g and from 0.02 to 0.27 nmol/g, respectively. The assay has been successfully applied to examine the FAEE levels in 81 meconium samples from babies born to mothers reporting alcohol consumption, to varying degrees, during pregnancy.
Sigma-Aldrich
Ethyl linoleate, ≥99%
Sigma-Aldrich
Ethyl linolenate, ≥98%