Merck
Beat-to-beat dynamic regulation of intracellular pH in cardiomyocytes.
iScience (2022-01-11)
Yankun Lyu, Phung N Thai, Lu Ren, Valeriy Timofeyev, Zhong Jian, Seojin Park, Kenneth S Ginsburg, James Overton, Julie Bossuyt, Donald M Bers, Ebenezer N Yamoah, Ye Chen-Izu, Nipavan Chiamvimonvat, Xiao-Dong Zhang
ABSTRACT
The mammalian heart beats incessantly with rhythmic mechanical activities generating acids that need to be buffered to maintain a stable intracellular pH (pHi) for normal cardiac function. Even though spatial pHi non-uniformity in cardiomyocytes has been documented, it remains unknown how pHi is regulated to match the dynamic cardiac contractions. Here, we demonstrated beat-to-beat intracellular acidification, termed pHi transients, in synchrony with cardiomyocyte contractions. The pHi transients are regulated by pacing rate, Cl-/HCO3 - transporters, pHi buffering capacity, and β-adrenergic signaling. Mitochondrial electron-transport chain inhibition attenuates the pHi transients, implicating mitochondrial activity in sculpting the pHi regulation. The pHi transients provide dynamic alterations of H+ transport required for ATP synthesis, and a decrease in pHi may serve as a negative feedback to cardiac contractions. Current findings dovetail with the prevailing three known dynamic systems, namely electrical, Ca2+, and mechanical systems, and may reveal broader features of pHi handling in excitable cells.
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Protease from Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder
Sigma-Aldrich
2,3-Butanedione monoxime, ≥98%
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
(−)-Isoproterenol hydrochloride
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amiloride