Merck
  • Synthesis and characterization of novel ssDNA X-aptamers targeting Growth Hormone Releasing Hormone (GHRH).
Synthesis and characterization of novel ssDNA X-aptamers targeting Growth Hormone Releasing Hormone (GHRH).
PloS one (2022-01-22)
Burcu Ayhan-Sahin, Zeynep-Elif Apaydın, Pınar Obakan-Yerlikaya, Elif-Damla Arisan, Ajda Coker-Gurkan
ABSTRACT
Growth Hormone Releasing Hormone (GHRH), 44 amino acids containing hypothalamic hormone, retains the biological activity by its first 29 amino acids. GHRH (NH2 1-29) peptide antagonists inhibit the growth of prostate, breast, ovarian, renal, gastric, pancreatic cancer in vitro and in vivo. Aptamers, single-strand RNA, or DNA oligonucleotides are capable of binding to target molecules with high affinity. Our aim in this study is to synthesize and select X-aptamers against both GHRH NH2 (1-29) and GHRH NH2 (1-44) and demonstrate synthesized aptamers' target binding activity as well as serum stability. Aptamers against GHRH NH2 (1-44) and NH2 (1-29) peptides were synthesized, and binding affinity (Kd) of 24 putative X-aptamers was determined by the dot-blot method, co-immunofluorescence staining and, SPR analysis. The serum stability of TKY.T1.08, TKY1.T1.13, TKY.T2.08, TKY.T2.09 X-aptamers was 90-120 h, respectively. The dose-dependent binding of TKY1.T1.13, TKY.T2.08, TKY.T2.09 X-aptamers on GHRHR in MIA PaCa-2 was approved by co-IF assay results. Moreover, SPR analysis indicated the Kd (4.75, 1.21, and 4.0 nM) levels of TKY2.T1.13, TKY.T2.08, TKY.T2.09 putative X-aptamers, respectively. Our results illustrate the synthesis of 24 putative X-aptamers against both GHRH NH2 (1-44) and NH2 (1-29) peptides and TKY1.T1.13, TKY.T2.08, TKY.T2.09 X-aptamers have high serum stability, high target binding potential with low Kd levels.
Sigma-Aldrich
Growth Hormone Releasing Factor Fragment 1-29 amide human, ≥97% (HPLC)
Pricing and availability is not currently available.