Merck
  • R7BP augments the function of RGS7*Gbeta5 complexes by a plasma membrane-targeting mechanism.
R7BP augments the function of RGS7*Gbeta5 complexes by a plasma membrane-targeting mechanism.
The Journal of biological chemistry (2006-07-27)
Ryan M Drenan, Craig A Doupnik, Muralidharan Jayaraman, Abigail L Buchwalter, Kevin M Kaltenbronn, James E Huettner, Maurine E Linder, Kendall J Blumer
ABSTRACT
The RGS7 (R7) family of G protein regulators, Gbeta5, and R7BP form heterotrimeric complexes that potently regulate the kinetics of G protein-coupled receptor signaling. Reversible palmitoylation of R7BP regulates plasma membrane/nuclear shuttling of R7*Gbeta5*R7BP heterotrimers. Here we have investigated mechanisms whereby R7BP controls the function of the R7 family. We show that unpalmitoylated R7BP undergoes nuclear/cytoplasmic shuttling and that a C-terminal polybasic motif proximal to the palmitoylation acceptor sites of R7BP mediates nuclear localization, palmitoylation, and plasma membrane targeting. These results suggest a novel mechanism whereby palmitoyltransferases and nuclear import receptors both utilize the C-terminal domain of R7BP to determine the trafficking fate of R7*Gbeta5*R7BP heterotrimers. Analogous mechanisms may regulate other signaling proteins whose distribution between the plasma membrane and nucleus is controlled by palmitoylation. Lastly, we show that cytoplasmic RGS7*Gbeta5*R7BP heterotrimers and RGS7*Gbeta5 heterodimers are equivalently inefficient regulators of G protein-coupled receptor signaling relative to plasma membrane-bound heterotrimers bearing palmitoylated R7BP. Therefore, R7BP augments the function of the complex by a palmitoylation-regulated plasma membrane-targeting mechanism.
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 9E10, clone 9E10, Upstate®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution