Merck
  • CRISPR screen identifies genes that sensitize AML cells to double-negative T-cell therapy.
CRISPR screen identifies genes that sensitize AML cells to double-negative T-cell therapy.
Blood (2020-12-04)
Fraser Soares, Branson Chen, Jong Bok Lee, Musaddeque Ahmed, Dalam Ly, Enoch Tin, Hyeonjeong Kang, Yong Zeng, Nayeema Akhtar, Mark D Minden, Housheng Hansen He, Li Zhang
ABSTRACT
Acute myeloid leukemia (AML) remains a devastating disease in need of new therapies to improve patient survival. Targeted adoptive T-cell therapies have achieved impressive clinical outcomes in some B-cell leukemias and lymphomas but not in AML. Double-negative T cells (DNTs) effectively kill blast cells from the majority of AML patients and are now being tested in clinical trials. However, AML blasts obtained from ∼30% of patients show resistance to DNT-mediated cytotoxicity; the markers or mechanisms underlying this resistance have not been elucidated. Here, we used a targeted clustered regularly interspaced short palindromic repeats (CRISPR)/CRISPR-associated protein 9 (Cas9) screen to identify genes that cause susceptibility of AML cells to DNT therapy. Inactivation of the Spt-Ada-Gcn5-acetyltransferase (SAGA) deubiquitinating complex components sensitized AML cells to DNT-mediated cytotoxicity. In contrast, CD64 inactivation resulted in resistance to DNT-mediated cytotoxicity. Importantly, the level of CD64 expression correlated strongly with the sensitivity of AML cells to DNT treatment. Furthermore, the ectopic expression of CD64 overcame AML resistance to DNTs in vitro and in vivo. Altogether, our data demonstrate the utility of CRISPR/Cas9 screens to uncover mechanisms underlying the sensitivity to DNT therapy and suggest CD64 as a predictive marker for response in AML patients.
Sigma-Aldrich
GenElute HP Plasmid Maxiprep Kit, sufficient for 25 purifications