Merck
  • Conformational analyses of 2,3-dihydroxypropanoic acid as a function of solvent and ionization state as determined by NMR spectroscopy.
Conformational analyses of 2,3-dihydroxypropanoic acid as a function of solvent and ionization state as determined by NMR spectroscopy.
Magnetic resonance in chemistry : MRC (2006-02-16)
Michael D Drake, Alex K Harsha, Sergei Terterov, John D Roberts
ABSTRACT
Vicinal (1)H--(1)H coupling constants were used to determine the conformational preferences of 2,3-dihydroxypropanoic acid (1) (DL-glyceric acid) in various solvents and its different carboxyl ionization states. The stereospecific assignments of J(12) and J(13) were confirmed through the point-group substitution of the C-3 hydrogen with deuterium, yielding rac-(2SR,3RS)-[3-(2)H]-1, and the observation of only J(13) in the (1)H NMR spectra. While hydrogen bonding and steric strain may be expected to drive the conformational equilibrium, their role is overshadowed by a profound gauche effect between the vicinal hydroxyl groups that mimics other substituted ethanes, such as 1,2-ethanediol and 1,2-difluoroethane. At low pH, the conformational equilibrium is heavily weighted toward the gauche-hydroxyl rotamers with a range of 81% in DMSO-d(6) to 92% in tert-butyl alcohol-d(10). At high pH, the equilibrium exhibits a larger dependence upon the polarity and solvating capability of the medium, although the gauche effect still dominates in D(2)O, 1,4-dioxane-d(8), methanol-d(4), and ethanol-d(6) (96, 89, 85, and 83% gauche-hydroxyls respectively). The observed preference for the gauche-hydroxyl rotamers is believed to stem primarily from hyperconjugative sigma(C--H) --> sigma*(C--OH) interactions.
Sigma-Aldrich
tert-Butanol-d10, 99 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Glyceric acid hemicalcium salt hydrate, ≥98% (GC)
Pricing and availability is not currently available.