Merck
  • Turn-on chemiluminescence probes and dual-amplification of signal for detection of amyloid beta species in vivo.
Turn-on chemiluminescence probes and dual-amplification of signal for detection of amyloid beta species in vivo.
Nature communications (2020-08-15)
Jing Yang, Wei Yin, Richard Van, Keyi Yin, Peng Wang, Chao Zheng, Biyue Zhu, Kathleen Ran, Can Zhang, Mohanraja Kumar, Yihan Shao, Chongzhao Ran
ABSTRACT
Turn-on fluorescence imaging is routinely studied; however, turn-on chemiluminescence has been rarely explored for in vivo imaging. Herein, we report the design and validation of chemiluminescence probe ADLumin-1 as a turn-on probe for amyloid beta (Aβ) species. Two-photon imaging indicates that ADLumin-1 can efficiently cross the blood-brain barrier and provides excellent contrast for Aβ plaques and cerebral amyloid angiopathy. In vivo brain imaging shows that the chemiluminescence signal of ADLumin-1 from 5-month-old transgenic 5xFAD mice is 1.80-fold higher than that from the age-matched wild-type mice. Moreover, we demonstrate that it is feasible to further dually-amplify signal via chemiluminescence resonance energy transfer (DAS-CRET) using two non-conjugated smart probes (ADLumin-1 and CRANAD-3) in solutions, brain homogenates, and in vivo whole brain imaging. Our results show that DAS-CRET can provide a 2.25-fold margin between 5-month-old 5xFAD mice and wild type mice. We believe that our strategy could be extended to other aggregating-prone proteins.
Sigma-Aldrich
CRANAD-3, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
ADLumin-1, ≥98% (HPLC)
Pricing and availability is not currently available.