Merck
  • Specific detection of Legionella pneumophila: construction of a new 16S rRNA-targeted oligonucleotide probe.
Specific detection of Legionella pneumophila: construction of a new 16S rRNA-targeted oligonucleotide probe.
Applied and environmental microbiology (1998-07-02)
D Grimm, H Merkert, W Ludwig, K H Schleifer, J Hacker, B C Brand
ABSTRACT
Based on comparative sequence analysis, we have designed an oligonucleotide probe complementary to a region of 16S rRNA of Legionella pneumophila which allows the differentiation of L. pneumophila from other Legionella species without cultivation. The specificity of the new probe, LEGPNE1, was tested by in situ hybridization to a total of four serogroups of six strains of L. pneumophila, five different Legionella spp. and three nonlegionella species as reference strains. Furthermore, L. pneumophila cells could be easily distinguished from Legionella micdadei and Pseudomonas aeruginosa cells by using in situ hybridization with probes LEGPNE1, LEG705, and EUB338 after infection of the protozoan Acanthamoeba castellanii.
Sigma-Aldrich
Escherichia coli FISH probe -ATTO488, Probe for fluorescence in situ hybridization (FISH), 20µM in water
Pricing and availability is not currently available.
Sigma-Aldrich
Escherichia coli FISH probe - Cy3, Probe for fluorescence in situ hybridization (FISH)
Pricing and availability is not currently available.