Merck
  • Mass production of human airway-like neutrophils via transmigration in an organotypic model of human airways.
Mass production of human airway-like neutrophils via transmigration in an organotypic model of human airways.
STAR protocols (2021-11-09)
Brian Dobosh, Vincent D Giacalone, Camilla Margaroli, Rabindra Tirouvanziam
ABSTRACT
Neutrophils are difficult to study, particularly in tissues, due to their short half-life and propensity for activation. We describe an organotypic airway model that uses patient airway fluid to enable the transmigration of blood neutrophils to acquire an airway-like phenotype in order to better understand their contribution to airway diseases. In particular, we showcase how conditioned neutrophils modulate their bacteria-killing abilities. For complete details on the use and execution of this protocol, please refer to Margaroli et al. (2021).
Sigma-Aldrich
4-Chloro-2-nitrobenzyl alcohol, 98%
Pricing and availability is not currently available.