Merck
  • Massive efflux of adenosine triphosphate into the extracellular space immediately after experimental traumatic brain injury.
Massive efflux of adenosine triphosphate into the extracellular space immediately after experimental traumatic brain injury.
Experimental and therapeutic medicine (2021-04-15)
Nobuhiro Moro, Sima S Ghavim, Richard L Sutton
ABSTRACT
The aim of the current study was to determine effects of mild traumatic brain injury (TBI), with or without blockade of purinergic ATP Y1 (P2Y1) receptors or store-operated calcium channels, on extracellular levels of ATP, glutamate, glucose and lactate. Concentrations of ATP, glutamate, glucose and lactate were measured in cerebral microdialysis samples obtained from the ipsilateral cortex and underlying hippocampus of rats with mild unilateral controlled cortical impact (CCI) or sham injury. Immediately after CCI, a large release of ATP was observed in the cortex (3.53-fold increase of pre-injury value) and hippocampus (2.97-fold increase of pre-injury value), with ATP returning to the baseline levels within 20 min post-injury and remaining stable for during the 3-h sampling period. In agreement with the results of previous studies, there was a significant increase in glutamate 20 min after CCI, which was concomitant with a decrease in extracellular glucose (20 min) and an increase in lactate (40-60 min) in both brain regions after CCI. Addition of a selective P2Y1 receptor blocker (MRS2179 ammonium salt hydrate) to the microdialysis perfusate significantly lowered pre-injury ATP and glutamate levels, and eliminated the post-CCI peaks. Addition of a blocker of store-operated calcium channels [2-aminoethoxy diphenylborinate (2-APB)] to the microdialysis perfusate significantly lowered pre-injury ATP in the hippocampus, and attenuated the post-CCI peak in both the cortex and hippocampus. 2-APB treatment significantly increased baseline glutamate levels, but the values post-injury did not differ from those in the sham group. Pre-injury glucose levels, but not lactate levels, were increased by MRS2179 and decreased by 2-APB. However, none of these treatments substantially altered the CCI-induced reduction in glucose and increase in lactate in the cortex. In conclusion, the results of the present study demonstrated that a short although extensive release of ATP immediately after experimental TBI can be significantly attenuated by blockade of P2Y1 receptors or store-operated calcium channels.
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) assay mix dilution buffer, lyophilized powder
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) assay mix, lyophilized powder
Sigma-Aldrich
MRS 2179 ammonium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%