Merck
  • Replacement of Volatile Acetic Acid by Solid SiO2@COOH Silica (Nano)Beads for (Ep)Oxidation Using Mn and Fe Complexes Containing BPMEN Ligand.
Replacement of Volatile Acetic Acid by Solid SiO2@COOH Silica (Nano)Beads for (Ep)Oxidation Using Mn and Fe Complexes Containing BPMEN Ligand.
Molecules (Basel, Switzerland) (2021-09-29)
Yun Wang, Florence Gayet, Jean-Claude Daran, Pascal Guillo, Dominique Agustin
ABSTRACT
Mn and Fe BPMEN complexes showed excellent reactivity in catalytic oxidation with an excess of co-reagent (CH3COOH). In the straight line of a cleaner catalytic system, volatile acetic acid was replaced by SiO2 (nano)particles with two different sizes to which pending carboxylic functions were added (SiO2@COOH). The SiO2@COOH beads were obtained by the functionalization of SiO2 with pending nitrile functions (SiO2@CN) followed by CN hydrolysis. All complexes and silica beads were characterized by NMR, infrared, DLS, TEM, X-ray diffraction. The replacement of CH3COOH by SiO2@COOH (100 times less on molar ratio) has been evaluated for (ep)oxidation on several substrates (cyclooctene, cyclohexene, cyclohexanol) and discussed in terms of activity and green metrics.
Sigma-Aldrich
4-Ethylmorpholine, ≥97.0% (GC)
Pricing and availability is not currently available.