Merck
  • Improvement of a synthetic live bacterial therapeutic for phenylketonuria with biosensor-enabled enzyme engineering.
Improvement of a synthetic live bacterial therapeutic for phenylketonuria with biosensor-enabled enzyme engineering.
Nature communications (2021-10-30)
Kristin J Adolfsen, Isolde Callihan, Catherine E Monahan, Per Jr Greisen, James Spoonamore, Munira Momin, Lauren E Fitch, Mary Joan Castillo, Lindong Weng, Lauren Renaud, Carl J Weile, Jay H Konieczka, Teodelinda Mirabella, Andres Abin-Fuentes, Adam G Lawrence, Vincent M Isabella
ABSTRACT
In phenylketonuria (PKU) patients, a genetic defect in the enzyme phenylalanine hydroxylase (PAH) leads to elevated systemic phenylalanine (Phe), which can result in severe neurological impairment. As a treatment for PKU, Escherichia coli Nissle (EcN) strain SYNB1618 was developed under Synlogic's Synthetic Biotic™ platform to degrade Phe from within the gastrointestinal (GI) tract. This clinical-stage engineered strain expresses the Phe-metabolizing enzyme phenylalanine ammonia lyase (PAL), catalyzing the deamination of Phe to the non-toxic product trans-cinnamate (TCA). In the present work, we generate a more potent EcN-based PKU strain through optimization of whole cell PAL activity, using biosensor-based high-throughput screening of mutant PAL libraries. A lead enzyme candidate from this screen is used in the construction of SYNB1934, a chromosomally integrated strain containing the additional Phe-metabolizing and biosafety features found in SYNB1618. Head-to-head, SYNB1934 demonstrates an approximate two-fold increase in in vivo PAL activity compared to SYNB1618.
Sigma-Aldrich
Hippuric acid, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
LB Broth with agar (Lennox), Highly-referenced microbial growth powder medium with Agar, low salt, suitable for salt-sensitive E. coli culture.
Pricing and availability is not currently available.