Merck
  • Targeting the interleukin-15/interleukin-15 receptor system in inflammatory autoimmune diseases.
Targeting the interleukin-15/interleukin-15 receptor system in inflammatory autoimmune diseases.
Arthritis research & therapy (2004-07-01)
Thomas A Waldmann
ABSTRACT
Interleukin (IL)-15 is a dangerous inflammatory cytokine that induces tumor-necrosis factor-alpha, IL-1beta and inflammatory chemokines. It inhibits self-tolerance mediated by IL-2 mediated activation-induced cell death and facilitates maintenance of CD8+ memory T-cell survival including that of self-directed memory cells. Disordered IL-15 expression has been reported in patients with an array of inflammatory autoimmune diseases. A series of therapeutic agents that inhibit IL-15 action have been introduced, including the soluble IL-15 receptor (IL-15R) alpha chain, mutant IL-15, and antibodies directed against the IL-15 cytokine and against the IL-2R/IL-15R beta subunit used by IL-2 and IL-15.
Sigma-Aldrich
Human IL-15 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Pricing and availability is not currently available.