Merck
  • A Potent Anti-Malarial Human Monoclonal Antibody Targets Circumsporozoite Protein Minor Repeats and Neutralizes Sporozoites in the Liver.
A Potent Anti-Malarial Human Monoclonal Antibody Targets Circumsporozoite Protein Minor Repeats and Neutralizes Sporozoites in the Liver.
Immunity (2020-09-19)
Lawrence T Wang, Lais S Pereira, Yevel Flores-Garcia, James O'Connor, Barbara J Flynn, Arne Schön, Nicholas K Hurlburt, Marlon Dillon, Annie S P Yang, Amanda Fabra-García, Azza H Idris, Bryan T Mayer, Monica W Gerber, Raphael Gottardo, Rosemarie D Mason, Nicole Cavett, Reid B Ballard, Neville K Kisalu, Alvaro Molina-Cruz, Jorgen Nelson, Rachel Vistein, Carolina Barillas-Mury, Rogerio Amino, David Baker, Neil P King, Robert W Sauerwein, Marie Pancera, Ian A Cockburn, Fidel Zavala, Joseph R Francica, Robert A Seder
ABSTRACT
Discovering potent human monoclonal antibodies (mAbs) targeting the Plasmodium falciparum circumsporozoite protein (PfCSP) on sporozoites (SPZ) and elucidating their mechanisms of neutralization will facilitate translation for passive prophylaxis and aid next-generation vaccine development. Here, we isolated a neutralizing human mAb, L9 that preferentially bound NVDP minor repeats of PfCSP with high affinity while cross-reacting with NANP major repeats. L9 was more potent than six published neutralizing human PfCSP mAbs at mediating protection against mosquito bite challenge in mice. Isothermal titration calorimetry and multiphoton microscopy showed that L9 and the other most protective mAbs bound PfCSP with two binding events and mediated protection by killing SPZ in the liver and by preventing their egress from sinusoids and traversal of hepatocytes. This study defines the subdominant PfCSP minor repeats as neutralizing epitopes, identifies an in vitro biophysical correlate of SPZ neutralization, and demonstrates that the liver is an important site for antibodies to prevent malaria.
Sigma-Aldrich
IGEPAL® CA-630, Molecular Biology
rProtein A Sepharose Fast Flow, Cytiva 17-1279-03, pack of 200 mL
Sigma-Aldrich
E-64, protease inhibitor