Merck
  • Membrane and raft association of reggie-1/flotillin-2: role of myristoylation, palmitoylation and oligomerization and induction of filopodia by overexpression.
Membrane and raft association of reggie-1/flotillin-2: role of myristoylation, palmitoylation and oligomerization and induction of filopodia by overexpression.
The Biochemical journal (2003-11-06)
Carolin Neumann-Giesen, Bianca Falkenbach, Peter Beicht, Stephanie Claasen, Georg Lüers, Claudia A O Stuermer, Volker Herzog, Ritva Tikkanen
ABSTRACT
The reggie protein family consists of two proteins, reggie-1 and -2, also called flotillins, which are highly ubiquitous and evolutionarily conserved. Both reggies have been shown to be associated with membrane rafts and are involved in various cellular processes such as T-cell activation, phagocytosis and insulin signalling. However, the exact molecular function of these proteins remains to be determined. In addition, the mechanism of membrane association of reggie-1, which does not contain any transmembrane domain, is not known. In this study, we have produced a fusion protein of reggie-1 with enhanced green fluorescent protein and generated targeted substitutions for the inactivation of putative palmitoylation and myristoylation sites. We were able to show that reggie-1 is myristoylated and multiply palmitoylated and that lipid modifications are necessary for membrane association of reggie-1. Overexpression of reggie-1 resulted in the induction of numerous filopodia-like protrusions in various cell lines, suggesting a role for reggie-1 as a signalling protein in actin-dependent processes.
Sigma-Aldrich
Escort IV Transfection Reagent, Lipid reagent for transient and stable transfection of mammalian and insect cells.