- Plant cell adhesion and growth on artificial fibrous scaffolds as an in vitro model for plant development.
Plant cell adhesion and growth on artificial fibrous scaffolds as an in vitro model for plant development.
Science advances (2021-10-21)
Ryan Calcutt, Richard Vincent, Derrick Dean, Treena Livingston Arinzeh, Ram Dixit
PMID34669469
ABSTRACT
[Figure: see text].
Product Description
Pricing and availability is not currently available.
Sigma-Aldrich
StableCell™ Trypsin Solution, 1X, sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Pricing and availability is not currently available.