Merck
Conversion of Escherichia coli to Generate All Biomass Carbon from CO2.
Cell (2019-11-30)
Shmuel Gleizer, Roee Ben-Nissan, Yinon M Bar-On, Niv Antonovsky, Elad Noor, Yehudit Zohar, Ghil Jona, Eyal Krieger, Melina Shamshoum, Arren Bar-Even, Ron Milo
ABSTRACT
The living world is largely divided into autotrophs that convert CO2 into biomass and heterotrophs that consume organic compounds. In spite of widespread interest in renewable energy storage and more sustainable food production, the engineering of industrially relevant heterotrophic model organisms to use CO2 as their sole carbon source has so far remained an outstanding challenge. Here, we report the achievement of this transformation on laboratory timescales. We constructed and evolved Escherichia coli to produce all its biomass carbon from CO2. Reducing power and energy, but not carbon, are supplied via the one-carbon molecule formate, which can be produced electrochemically. Rubisco and phosphoribulokinase were co-expressed with formate dehydrogenase to enable CO2 fixation and reduction via the Calvin-Benson-Bassham cycle. Autotrophic growth was achieved following several months of continuous laboratory evolution in a chemostat under intensifying organic carbon limitation and confirmed via isotopic labeling.
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Pricing and availability is not currently available.