Merck
Molecular Determinant of DIDS Analogs Targeting RAD51 Activity.
Molecules (Basel, Switzerland) (2021-09-29)
Denis Velic, Alexandre Demeyer, Thibaut Peterlini, Houda Benhelli-Mokrani, Monique Mathé-Allainmat, Jean-Yves Masson, Fabrice Fleury
ABSTRACT
RAD51 is the central protein in DNA repair by homologous recombination (HR), involved in several steps of this process. It is shown that overexpression of the RAD51 protein is correlated with increased survival of cancer cells to cancer treatments. For the past decade, RAD51 overexpression-mediated resistance has justified the development of targeted inhibitors. One of the first molecules described to inhibit RAD51 was the 4,4'-diisothiocyanato-stilbene-2,2'-disulfonic acid (DIDS) molecule. This small molecule is effective in inhibiting different functions of RAD51, however its mode of action and the chemical functions involved in this inhibition have not been identified. In this work, we used several commercial molecules derived from DIDS to characterize the structural determinants involved in modulating the activity of RAD51. By combining biochemical and biophysical approaches, we have shown that DIDS and two analogs were able to inhibit the binding of RAD51 to ssDNA and prevent the formation of D-loop by RAD51. Both isothiocyanate substituents of DIDS appear to be essential in the inhibition of RAD51. These results open the way to the synthesis of new molecules derived from DIDS that should be greater modulators of RAD51 and more efficient for HR inhibition.
Sigma-Aldrich
Anti-RAD51 antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.