Merck
  • Loss of N1-methylation of G37 in tRNA induces ribosome stalling and reprograms gene expression.
Loss of N1-methylation of G37 in tRNA induces ribosome stalling and reprograms gene expression.
eLife (2021-08-13)
Isao Masuda, Jae-Yeon Hwang, Thomas Christian, Sunita Maharjan, Fuad Mohammad, Howard Gamper, Allen R Buskirk, Ya-Ming Hou
ABSTRACT
N1-methylation of G37 is required for a subset of tRNAs to maintain the translational reading-frame. While loss of m1G37 increases ribosomal +1 frameshifting, whether it incurs additional translational defects is unknown. Here, we address this question by applying ribosome profiling to gain a genome-wide view of the effects of m1G37 deficiency on protein synthesis. Using E coli as a model, we show that m1G37 deficiency induces ribosome stalling at codons that are normally translated by m1G37-containing tRNAs. Stalling occurs during decoding of affected codons at the ribosomal A site, indicating a distinct mechanism than that of +1 frameshifting, which occurs after the affected codons leave the A site. Enzyme- and cell-based assays show that m1G37 deficiency reduces tRNA aminoacylation and in some cases peptide-bond formation. We observe changes of gene expression in m1G37 deficiency similar to those in the stringent response that is typically induced by deficiency of amino acids. This work demonstrates a previously unrecognized function of m1G37 that emphasizes its role throughout the entire elongation cycle of protein synthesis, providing new insight into its essentiality for bacterial growth and survival.
Product Description
Roche
Nuclease S7, Micrococcal nuclease, from Staphylococcus aureus
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody