Merck
  • Low expression of the PPARγ-regulated gene thioredoxin-interacting protein accompanies human melanoma progression and promotes experimental lung metastases.
Low expression of the PPARγ-regulated gene thioredoxin-interacting protein accompanies human melanoma progression and promotes experimental lung metastases.
Scientific reports (2021-04-14)
Patrick Meylan, Christine Pich, Carine Winkler, Stefanie Ginster, Lionel Mury, Marie Sgandurra, René Dreos, Dennie Tompers Frederick, Marc Hammond, Genevieve Marie Boland, Liliane Michalik
ABSTRACT
The thioredoxin system plays key roles in regulating cancer cell malignancy. Here we identify the Thioredoxin-interacting protein (TXNIP) as a gene, which expression is regulated by PPARγ in melanoma cells. We show that high TXNIP expression levels associate with benign melanocytic lesions, with tumor regression in patients on MAP kinase targeted therapy, with decreased proliferation in patients' melanoma biopsies, and with cell cycle arrest in human melanoma cell lines. In contrast, reduced TXNIP expression associates with advanced melanoma and with disease progression in patients. TXNIP depletion in human melanoma cells altered the expression of integrin beta-3 and the localization of the integrin alpha-v/beta-3 dimer at their surface. Moreover, TXNIP depletion affected human melanoma cell motility and improved their capacity to colonize mouse lungs in an in vivo assay. This study establishes TXNIP as a PPARγ-regulated gene in melanoma cells, thereby suggesting a link between these two proteins both involved in the regulation of cancer and of energy metabolism. It also reveals that the decrease in TXNIP expression, which is observed in advanced patient tumors, likely favors lung metastatic seeding of malignant cells.
Sigma-Aldrich
Anti-MMP-1 (Ab-1) Mouse mAb (41-1E5), liquid, clone 41-1E5, Calbiochem®