Merck
  • The collagen structure of C1q induces wound healing by engaging discoidin domain receptor 2.
The collagen structure of C1q induces wound healing by engaging discoidin domain receptor 2.
Molecular medicine (Cambridge, Mass.) (2021-10-05)
Ria Aryani Hayuningtyas, Myeonggil Han, Seoyeon Choi, Man Sup Kwak, In Ho Park, Ji-Hyun Lee, Ji Eun Choi, Dae Ki Kim, Myoungsun Son, Jeon-Soo Shin
ABSTRACT
C1q has been reported to reveal complement-independent roles in immune and non-immune cells. C1q binds to its specific receptors to regulate distinct functions that rely on the environment and cell types. Discoidin domain receptor 2 (DDR2) is activated by collagen and functions in wound healing by controlling matrix metalloproteinase (MMP) expression. Since C1q exhibits a collagen-like structure, we hypothesized that C1q might engage DDR2 to regulate wound healing and extracellular matrix (ECM) remodeling. Cell-based assay, proximity ligation assay, ELISA, and surface plasmon analysis were utilized to investigate DDR2 and C1q binding. We also investigate the C1q-mediated in vitro wound healing ability using the human fibrosarcoma cell line, HT1080. C1q induced the phosphorylation of DDR2, p38 kinase, and ERK1/2. C1q and DDR2 binding improved cell migration and induced MMP2 and MMP9 expression. DDR2-specific shRNA reduced C1q-mediated cell migration for wound healing. C1q is a new DDR2 ligand that promotes wound healing. These findings have therapeutic implications in wound healing-related diseases.
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Complement component C1q from human serum, ≥95% (SDS-PAGE)
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Pricing and availability is not currently available.