Merck
  • MUC1-C influences cell survival in lung adenocarcinoma Calu-3 cells after SARS-CoV-2 infection.
MUC1-C influences cell survival in lung adenocarcinoma Calu-3 cells after SARS-CoV-2 infection.
BMB reports (2021-04-10)
Dongbum Kim, Sony Maharjan, Jinsoo Kim, Sangkyu Park, Jeong-A Park, Byoung Kwon Park, Younghee Lee, Hyung-Joo Kwon
ABSTRACT
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) induces coronavirus disease 2019 (COVID-19) and may increase the risk of adverse outcomes in lung cancer patients. In this study, we investigated the expression and function of mucin 1 (MUC1) after SARS-CoV-2 infection in the lung epithelial cancer cell line Calu-3. MUC1 is a major constituent of the mucus layer in the respiratory tract and contributes to pathogen defense. SARS-CoV-2 infection induced MUC1 C-terminal subunit (MUC1-C) expression in a STAT3 activation-dependent manner. Inhibition of MUC1-C signaling increased apoptosis-related protein levels and reduced proliferation-related protein levels; however, SARS-CoV-2 replication was not affected. Together, these results suggest that increased MUC1-C expression in response to SARS-CoV-2 infection may trigger the growth of lung cancer cells, and COVID-19 may be a risk factor for lung cancer patients. [BMB Reports 2021; 54(8): 425-430].
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid