Merck
  • Prognostic Significance of the Microbiome and Stromal Cells Phenotype in Esophagus Squamous Cell Carcinoma.
Prognostic Significance of the Microbiome and Stromal Cells Phenotype in Esophagus Squamous Cell Carcinoma.
Biomedicines (2021-07-03)
Olga Kovaleva, Polina Podlesnaya, Madina Rashidova, Daria Samoilova, Anatoly Petrenko, Valeria Mochalnikova, Vladimir Kataev, Yuri Khlopko, Andrey Plotnikov, Alexei Gratchev
ABSTRACT
Esophageal cancer is one of the most aggressive malignant neoplasms, with low survival rates and limited treatment options. In this study we analyzed the microbiome composition and the phenotype of inflammatory tumor infiltrate in squamous cell carcinoma of esophagus (ESCC) and examined possible relationships between them and their prognostic significance. We found that the predominant phyla of microorganisms found in both tumors and adjacent normal tissues were Firmicutes, Proteobacteria, Actinobacteria, Gemmatimonadetes and Bacteroidetes. We established that only bacteria of the genus Staphylococcus differ between tumors and normal tissues. We found a significant correlation between bacterial burden and the phenotype of the tumor stroma. Namely, a group of tumors characterized by a high expression of CD206 (r = -0.3976, p = 0.0056) in the stroma and iNOS (r = -0.2953, p = 0.0439) in tumor cells is characterized by a higher bacterial burden. Further, we established that in the group with a high content of CD206+ macrophages, there is also a predominance of gram-positive bacteria over gram-negative ones. We found that gram-positive bacterial burden is associated with disease prognosis in ESCC showing high content of CD206+ macrophages. In conclusion we established that the tumor microbiome, can be prognostically significant for ESCC when combined with other stromal markers.
Sigma-Aldrich
Anti-MRC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-INOS antibody, Rabbit monoclonal, clone SP126, recombinant, expressed in proprietary host, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-MSR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.