Merck
  • Overexpression of mutant amyloid-β protein precursor and presenilin 1 modulates enteric nervous system.
Overexpression of mutant amyloid-β protein precursor and presenilin 1 modulates enteric nervous system.
Journal of Alzheimer's disease : JAD (2014-11-20)
Kendra L Puig, Brianna M Lutz, Siri A Urquhart, Andrew A Rebel, Xudong Zhou, Gunjan D Manocha, MaryAnn Sens, Ashok K Tuteja, Norman L Foster, Colin K Combs
ABSTRACT
Alzheimer's disease (AD) is a neurodegenerative disorder histologically characterized by amyloid-β (Aβ) protein accumulation and activation of associated microglia. Although these features are well described in the central nervous system, the process and consequences of Aβ accumulation in the enteric nervous system have not been extensively studied. We hypothesized that Aβ also may accumulate in the enteric nervous system and lead to immune cell activation and neuronal dysfunction in the digestive tract not unlike that observed in diseased brain. To test this hypothesis, ileums of the small intestine of thirteen month old AβPP/PS1 and C57BL/6 (wild type) mice were collected and analyzed using immunohistochemistry, western blot analysis, cytokine arrays, and ELISA. AβPP/PS1 mice demonstrated no differences in intestinal motility or water absorption but elevated luminal IgA levels compared to wild type mice. They also had increased protein levels of AβPP and the proteolytic enzyme, BACE, corresponding to an increase in Aβ1-40 in the intestinal lysate as well as an increase in both Aβ1-40 and Aβ1-42 in the stool. This correlated with increased protein markers of proinflammatory and immune cell activation. Histologic analysis localized AβPP within enteric neurons but also intestinal epithelial cells with elevated Aβ immunoreactivity in the AβPP/PS1 mice. The presence of AβPP, Aβ, and CD68 immunoreactivity in the intestines of some patients with neuropathologically-confirmed AD are consistent with the findings in this mouse model. These data support the hypothesis that in AD the intestine, much like the brain, may develop proinflammatory and immune changes related to AβPP and Aβ.
Product Description
Millipore
High Sensitivity Human Amyloid β40 ELISA, This High Sensitivity Human Amyloid β40 ELISA is used to measure & quantify Amyloid β40 levels in Neuroscience research.
Pricing and availability is not currently available.
Millipore
High Sensitivity Human Amyloid β42 ELISA, This High Sensitivity Human Amyloid β42 ELISA is used to measure & quantify Amyloid β42 levels in Neuroscience research.
Pricing and availability is not currently available.