Merck
  • A Chaperone Lid Ensures Efficient and Privileged Client Transfer during Tail-Anchored Protein Targeting.
A Chaperone Lid Ensures Efficient and Privileged Client Transfer during Tail-Anchored Protein Targeting.
Cell reports (2019-01-04)
Un Seng Chio, SangYoon Chung, Shimon Weiss, Shu-Ou Shan
ABSTRACT
Molecular chaperones play key roles in maintaining cellular proteostasis. In addition to preventing client aggregation, chaperones often relay substrates within a network while preventing off-pathway chaperones from accessing the substrate. Here we show that a conserved lid motif lining the substrate-binding groove of the Get3 ATPase enables these important functions during the targeted delivery of tail-anchored membrane proteins (TAs) to the endoplasmic reticulum. The lid prevents promiscuous TA handoff to off-pathway chaperones, and more importantly, it cooperates with the Get4/5 scaffolding complex to enable rapid and privileged TA transfer from the upstream co-chaperone Sgt2 to Get3. These findings provide a molecular mechanism by which chaperones maintain the pathway specificity of client proteins in the crowded cytosolic environment.
Sigma-Aldrich
Calcium acetate hydrate, ReagentPlus®, ≥99% (titration), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Adenosine 5′-diphosphate sodium salt, bacterial, ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
Pricing and availability is not currently available.
Sigma-Aldrich
Coenzyme A trilithium salt, ≥93%
Pricing and availability is not currently available.