Merck
  • Methotrexate enhances antigen presentation and maturation of tumour antigen-loaded dendritic cells through NLRP3 inflammasome activation: a strategy for dendritic cell-based cancer vaccine.
Methotrexate enhances antigen presentation and maturation of tumour antigen-loaded dendritic cells through NLRP3 inflammasome activation: a strategy for dendritic cell-based cancer vaccine.
Therapeutic advances in medical oncology (2021-02-23)
Gao-Na Shi, Min Hu, Chengjuan Chen, Junmin Fu, Shuai Shao, Yu Zhou, Lei Wu, Tiantai Zhang
ABSTRACT
Dendritic cells (DCs) are antigen-presenting cells that play a pivotal role in adaptive cell-mediated immunity by priming and activating T cells against specific tumour and pathogenic antigens. Methotrexate (MTX), a folate derivative, functions as an immunoregulatory agent. However, the possible effect of MTX on tumour antigen-loaded DCs has not yet been investigated. We analysed the effect of MTX on the maturation and function of DCs along with tumour cell lysates (TCLs). Using bone marrow-derived DCs, we investigated the effect of MTX combined TCL-loaded DCs on T cells priming and proliferation. We also tested the anti-tumour immune effect on DCs when treated with MTX and/or TCL in vivo. MTX combined with TCL not only enhanced DC maturation and stimulated cytokine release but also promoted CD8+ T cell activation and proliferation. The latter was associated with increased tumour antigen uptake and cross-presentation to T cells. Mechanistically, DC maturation and antigen presentation were partly modulated by NLRP3 inflammasome activation. Furthermore, immunisation of mice with MTX and TCL-pulsed DCs before a tumour challenge significantly delayed tumour onset and retarded its growth. This protective effect was due to priming of IFN-γ releasing CD8+ T cells and enhanced killing of tumour cells by cytotoxic T lymphocytes isolated from these immunised mice. MTX can function as a potent adjuvant in DC vaccines by increasing antigen presentation and T cell priming. Our findings provide a new strategy for the application of DC-based anti-tumour immunotherapy.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, tested according to Ph. Eur.