Merck
  • Critical Role of Dipeptidyl Peptidase IV: A Therapeutic Target for Diabetes and Cancer.
Critical Role of Dipeptidyl Peptidase IV: A Therapeutic Target for Diabetes and Cancer.
Mini reviews in medicinal chemistry (2018-04-26)
Sourav De, Subhasis Banerjee, S K Ashok Kumar, Priyankar Paira
ABSTRACT
Diabetes mellitus is an emerging predator and affecting around 422 million adults worldwide. Higher levels of circulating insulin and increased pressure on the pancreas to produce insulin have been inferred as possible etiology for diabetes leading to a higher risk of pancreatic cancer. Out of several drug targets in hypoglycemic discovery, Dipeptidyl peptidase-IV (DPP-IV) has been considered an emerging target. It is a protease enzyme which inactivates incretin hormones i.e., Glucagonlike peptide 1 (GLP-1) and glucose-dependent insulin tropic polypeptide (GIP). Inhibition of DPP-4 results in the longer action of GLP-1 and GIP, therefore, DPP-4 inhibitors play an important role in maintaining glucose homeostasis. In comparison to early oral hypoglycemic, DPP-IV inhibitors are well tolerated and provide a better glycemic control over a longer period. These enzymes are expressed in a dimeric form on the surface of different cells such as prostate, liver and small intestinal epithelium cells. Disruption of the local signaling environment is an emerging factor in cancer development. Till date, not even a single DPP-IV inhibitor as anticancer has been developed. This review focuses on various features of the enzyme and their suitable inhibitors for target disease.
Sigma-Aldrich
Dipeptidyl Peptidase IV Inhibitor I
Pricing and availability is not currently available.
Sigma-Aldrich
DPP IV Inhibitor
Pricing and availability is not currently available.