Merck
  • Nicotinamide N-Methyltransferase in Acquisition of Stem Cell Properties and Therapy Resistance in Cancer.
Nicotinamide N-Methyltransferase in Acquisition of Stem Cell Properties and Therapy Resistance in Cancer.
International journal of molecular sciences (2021-06-03)
Renata Novak Kujundžić, Marin Prpić, Nikola Đaković, Nina Dabelić, Marko Tomljanović, Anamarija Mojzeš, Ana Fröbe, Koraljka Gall Trošelj
ABSTRACT
The activity of nicotinamide N-methyltransferase (NNMT) is tightly linked to the maintenance of the nicotinamide adenine dinucleotide (NAD+) level. This enzyme catalyzes methylation of nicotinamide (NAM) into methyl nicotinamide (MNAM), which is either excreted or further metabolized to N1-methyl-2-pyridone-5-carboxamide (2-PY) and H2O2. Enzymatic activity of NNMT is important for the prevention of NAM-mediated inhibition of NAD+-consuming enzymes poly-adenosine -diphosphate (ADP), ribose polymerases (PARPs), and sirtuins (SIRTs). Inappropriately high expression and activity of NNMT, commonly present in various types of cancer, has the potential to disrupt NAD+ homeostasis and cellular methylation potential. Largely overlooked, in the context of cancer, is the inhibitory effect of 2-PY on PARP-1 activity, which abrogates NNMT's positive effect on cellular NAD+ flux by stalling liberation of NAM and reducing NAD+ synthesis in the salvage pathway. This review describes, and discusses, the mechanisms by which NNMT promotes NAD+ depletion and epigenetic reprogramming, leading to the development of metabolic plasticity, evasion of a major tumor suppressive process of cellular senescence, and acquisition of stem cell properties. All these phenomena are related to therapy resistance and worse clinical outcomes.
Sigma-Aldrich
N-Methyl-2-pyridone-5-carboxamide
Pricing and availability is not currently available.
Sigma-Aldrich
N-Methyl-4-pyridone-5-carboxamide
Pricing and availability is not currently available.