Merck
  • AKT3-mediated IWS1 phosphorylation promotes the proliferation of EGFR-mutant lung adenocarcinomas through cell cycle-regulated U2AF2 RNA splicing.
AKT3-mediated IWS1 phosphorylation promotes the proliferation of EGFR-mutant lung adenocarcinomas through cell cycle-regulated U2AF2 RNA splicing.
Nature communications (2021-08-01)
Georgios I Laliotis, Evangelia Chavdoula, Maria D Paraskevopoulou, Abdul Kaba, Alessandro La Ferlita, Satishkumar Singh, Vollter Anastas, Keith A Nair, Arturo Orlacchio, Vasiliki Taraslia, Ioannis Vlachos, Marina Capece, Artemis Hatzigeorgiou, Dario Palmieri, Christos Tsatsanis, Salvatore Alaimo, Lalit Sehgal, David P Carbone, Vincenzo Coppola, Philip N Tsichlis
ABSTRACT
AKT-phosphorylated IWS1 regulates alternative RNA splicing via a pathway that is active in lung cancer. RNA-seq studies in lung adenocarcinoma cells lacking phosphorylated IWS1, identified a exon 2-deficient U2AF2 splice variant. Here, we show that exon 2 inclusion in the U2AF2 mRNA is a cell cycle-dependent process that is regulated by LEDGF/SRSF1 splicing complexes, whose assembly is controlled by the IWS1 phosphorylation-dependent deposition of histone H3K36me3 marks in the body of target genes. The exon 2-deficient U2AF2 mRNA encodes a Serine-Arginine-Rich (RS) domain-deficient U2AF65, which is defective in CDCA5 pre-mRNA processing. This results in downregulation of the CDCA5-encoded protein Sororin, a phosphorylation target and regulator of ERK, G2/M arrest and impaired cell proliferation and tumor growth. Analysis of human lung adenocarcinomas, confirmed activation of the pathway in EGFR-mutant tumors and showed that pathway activity correlates with tumor stage, histologic grade, metastasis, relapse after treatment, and poor prognosis.
Sigma-Aldrich
Citrate Buffer, pH 6.0, 10×, Antigen Retriever, for immunohistochemistry
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Hexadimethrine bromide, ≥95%
Sigma-Aldrich
Guanidine hydrochloride solution, 6M