Merck
  • Chemotherapeutic Tumor Microparticles Elicit a Neutrophil Response Targeting Malignant Pleural Effusions.
Chemotherapeutic Tumor Microparticles Elicit a Neutrophil Response Targeting Malignant Pleural Effusions.
Cancer immunology research (2020-07-15)
Pingwei Xu, Ke Tang, Jingwei Ma, Huafeng Zhang, Dianheng Wang, Liyan Zhu, Jie Chen, Keke Wei, Jincheng Liu, Haiqing Fang, Liang Tang, Yi Zhang, Jing Xie, Yuying Liu, Rui Meng, Li Liu, Xiaorong Dong, Kunyu Yang, Gang Wu, Fei Ma, Bo Huang
ABSTRACT
Malignant pleural effusion (MPE) is a frequent complication of various cancers and often leads to a poor quality of life, prognosis, and life expectancy, and its management remains palliative. New approaches that can effectively treat MPE are highly desirable. Here, we show that methotrexate (MTX)-packaging tumor cell-derived microparticles (MTX-MP) act as an effective immunotherapeutic agent to treat patients with MPE by mobilizing and activating neutrophils. We find that MTX-MP perfusion via a pleural catheter elicits the recruitment of neutrophils in patients through macrophage-released CXCL1 and CXCL2. By performing ex vivo experiments, we find that the recruited neutrophils are activated and release reactive oxygen species (ROS) and neutrophil extracellular trap (NET) to kill tumor cells. Neutrophil-released NETs were also able to seal off the damaged endothelium, facilitating MPE resolution in vitro and in tumor-bearing mice. These findings reveal the potential for use of cell-derived materials to package drugs as an immunotherapeutic agent against MPE.
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Evans Blue, Dye content ≥75 %
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Mini Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 40,000
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%
Sigma-Aldrich
N-acetylcysteine amide, ≥98% (HPLC)
Sigma-Aldrich
Hoechst 33258 solution, 1 mg/mL in H2O, ≥98.0% (HPLC)
Sigma-Aldrich
Ionomycin calcium salt, Ready Made Solution, from Streptomyces conglobatus, 1 mM in DMSO
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free