Merck
  • Histone H2B ubiquitination mediated chromatin relaxation is essential for the induction of somatic cell reprogramming.
Histone H2B ubiquitination mediated chromatin relaxation is essential for the induction of somatic cell reprogramming.
Cell proliferation (2021-06-23)
Liying Wang, Zhiliang Xu, Libin Wang, Chao Liu, Huafang Wei, Ruidan Zhang, Yinghong Chen, Lina Wang, Wenwen Liu, Sai Xiao, Wei Li, Wei Li
ABSTRACT
Cell reprogramming has significant impacts on their potential application in regenerative medicine. Chromatin remodelling plays a very important role in cell reprogramming, but its underlying mechanism remains poorly understood. RNA-seq, quantitative RT-PCR and western blot analysis were applied to study the role of RNF20 and H2B ubiquitination during mouse somatic cell reprogramming. Chromatin structure and the recruitment of transcription factors were analysed by ChIP-seq, micrococcal nuclease sensitivity assays and immunofluorescence staining. We show that RNF20 is highly expressed at the early stage of reprogramming along with the accumulation of H2B ubiquitination at the same stage, and Rnf20 knockout results in the failure of reprogramming at the initial stage but not the maturation and stabilization stages. RNA-seq showed that Rnf20 knockout mainly affects the early stage of cell reprogramming by impairing the transcription of MET-related genes and early pluripotency genes. Importantly, Rnf20 knockout results in a more compacted chromosomes structure in reprogrammable cells, suppressing the recruitment of reprogramming transcription factors to their proper locations on the chromosomes, and finally resulting in the failure of pluripotent gene network establishment. Histone H2B ubiquitination mediated chromatin relaxation is essential for the induction of somatic cell reprogramming.
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^7 units/ml.
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks