Merck
  • A caged E3 ligase ligand for PROTAC-mediated protein degradation with light.
A caged E3 ligase ligand for PROTAC-mediated protein degradation with light.
Chemical communications (Cambridge, England) (2020-04-17)
Cyrille S Kounde, Maria M Shchepinova, Charlie N Saunders, Marcel Muelbaier, Mark D Rackham, John D Harling, Edward W Tate
ABSTRACT
With the intent of achieving greater spatiotemporal control of PROTAC-induced protein degradation, a light-activated degrader was designed by photocaging an essential E3 ligase binding motif in a BRD4 targeting PROTAC. Proteolysis was triggered only after a short irradiation time, the kinetics of which could be monitored by live-cell video microscopy.
Sigma-Aldrich
(S,R,S)-AHPC-Me, 95%