Merck
  • Recombinant N-glycosylation isoforms of Legume lectins: Production and purification from Nicotiana benthamiana leaves following RuBisCO depletion.
Recombinant N-glycosylation isoforms of Legume lectins: Production and purification from Nicotiana benthamiana leaves following RuBisCO depletion.
Plant physiology and biochemistry : PPB (2020-11-20)
Kevin Bellande, Alexandre Lalo, Lætitia Ligat, David Roujol, Elisabeth Jamet, Hervé Canut
ABSTRACT
An efficient purification of recombinant proteins often requires a high ratio of recombinant to host proteins. In plants, Ribulose-1,5-bisphosphate carboxylase/oxygenase (RuBisCO) is the most abundant leaf protein, thus strongly impacting purification yield. Here, we describe a simple and robust purification procedure for recombinant proteins based on a differential precipitation of RuBisCO. In this context, four Legume lectin domains of Arabidopsis thaliana which belong to receptor-like kinases and cell wall proteins were produced from Nicotiana benthamiana leaves. The recombinant proteins exhibit a unique lectin domain consisting of around 250 amino acid residues with several predicted N-glycosylation sites and a six His-tag at the N-terminus. After ammonium sulphate precipitation of total soluble proteins, depletion of RuBisCO was obtained using citrate and succinate buffers during the salting-in step: this depletion was pH-dependent and the presence of di- or tri-carboxylic acids was required. The depleted protein extracts were then subjected to two chromatographic steps which were used in the negative mode to submit a protein fraction enriched as much as possible in recombinant lectin domains to a third chromatographic step (immobilized metal-ion chromatography). Three of the Legume lectin domains were purified near to homogeneity and revealed multiple N-glycosylation isoforms, particularly those from receptor-like kinases, which were characterised using specific lectins and deglycosylation enzymes. The production and purification of recombinant lectin domains will facilitate their biochemical characterisation in the context of cell-to-cell signalling and cell wall organisation.
Sigma-Aldrich
Protamine sulfate salt from salmon, Grade X, amorphous powder
Pricing and availability is not currently available.
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available.
Sigma-Aldrich
Protease Inhibitor Cocktail, for plant cell and tissue extracts, DMSO solution
Pricing and availability is not currently available.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethyleneimine) solution, average Mn ~60,000 by GPC, average Mw ~750,000 by LS, 50 wt. % in H2O
Pricing and availability is not currently available.