Merck
  • Stroke-Induced Peripheral Immune Dysfunction in Vitamin D-Deficient Conditions: Modulation by Progesterone and Vitamin D.
Stroke-Induced Peripheral Immune Dysfunction in Vitamin D-Deficient Conditions: Modulation by Progesterone and Vitamin D.
Molecular neurobiology (2020-10-17)
Seema Yousuf, Fahim Atif, Claudia Espinosa-Garcia, Wayne Harris, Nefize Turan, Donald G Stein
ABSTRACT
Vitamin D deficiency (Ddef) alters morphology and outcomes after a stroke. We investigated the interaction of Ddef following post-stroke systemic inflammation and evaluated whether administration of progesterone (P) or vitamin D (D) will improve outcomes. Ddef rats underwent stroke with lipopolysaccharide (LPS)-induced systemic inflammation. Rats were randomly divided into 9 groups and treated with P, D, or vehicle for 4 days. At day 4, rats were tested on different behavioral parameters. Markers of neuronal inflammation, endoplasmic reticulum stress, oxidative stress, white matter integrity, and apoptosis were measured along with immune cell populations from the spleen, thymus, and blood. Severely altered outcomes were observed in the Ddef group compared to the D-sufficient (Dsuf) group. Stroke caused peripheral immune dysfunction in the Dsuf group which was worse in the Ddef group. Systemic inflammation exacerbated injury outcomes in the Dsuf group and these were worse in the Ddef group. Monotherapy with P/D showed beneficial functional effects but the combined treatment showed better outcomes than either alone. Ddef as a comorbid condition with stroke worsens stroke outcomes and can delay functional recovery. Combination treatment with P and D might be promising for future stroke therapeutics in Ddef.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, Ready Made solution, 1 mg/mL, 0.2 μm filtered
Pricing and availability is not currently available.