Merck
  • Development of Artificial Urine Certified Reference Material for Quantification of Neonicotinoid Insecticides.
Development of Artificial Urine Certified Reference Material for Quantification of Neonicotinoid Insecticides.
Journal of AOAC International (2020-11-29)
Takamitsu Otake, Shoji F Nakayama, Tomohiko Isobe, Nobuyasu Hanari
ABSTRACT
Neonicotinoid insecticides (neonicotinoids) are widely used, however, they can negatively affect human health. Thus, neonicotinoids and their metabolites in human urine are analyzed globally to assess exposure levels and health risks. To properly assess the exposure levels and health risks, accurate analytical results are a necessity. This study aims to develop urine certified reference material (CRM), NMIJ CRM 7408-a, for the quantification of neonicotinoids. To develop NMIJ CRM 7408-a, commercially available artificial urine was used as the raw material to which the target neonicotinoids were added. Analyses of neonicotinoids in NMIJ CRM 7408-a were carried out by isotope dilution mass spectrometry (IDMS). Sample homogeneity was assessed, with the results showing that acetamiprid, clothianidin, thiacloprid, and thiamethoxam exhibited sufficiently homogeneous distributions in the material. The relative uncertainties due to inhomogeneity were 1.2-7.0%. The results obtained from long-term stability assessment indicated that the target neonicotinoids were stable. The relative uncertainties due to instability were 9.4-17.2% (for an expiry date of 21 months). The characterization for providing concentration values was carried out using one (clothianidin and thiacloprid) or two (acetamiprid and thiamethoxam) analytical methods. The certified values of the target neonicotinoids (acetamiprid, clothianidin, thiacloprid, and thiamethoxam) were 0.19-1.38 µg/kg. This is the first frozen artificial urine CRM in which neonicotinoids were quantified by IDMS and will be useful for evaluating the accuracy of analysis and validation of analytical methods for the determination of neonicotinoids in urine. NMIJ CRM 7408-a, which has been certified for four neonicotinoids, was developed by NMIJ.
Product Description
Supelco
Acetamiprid-N-desmethyl, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Fenobucarb, PESTANAL®, analytical standard
Pricing and availability is not currently available.