Merck
  • Synthesis and biological evaluation of novel irreversible serine protease inhibitors using amino acid based sulfonyl fluorides as an electrophilic trap.
Synthesis and biological evaluation of novel irreversible serine protease inhibitors using amino acid based sulfonyl fluorides as an electrophilic trap.
Bioorganic & medicinal chemistry (2011-03-23)
Arwin J Brouwer, Tarik Ceylan, Anika M Jonker, Tima van der Linden, Rob M J Liskamp
ABSTRACT
We have designed and synthesized novel irreversible serine protease inhibitors containing aliphatic sulfonyl fluorides as an electrophilic trap. These substituted taurine sulfonyl fluorides derived from taurine or protected amino acids were conveniently synthesized from β-aminoethanesulfonyl chlorides using KF/18-crown-6 or from β-aminoethanesulfonates using DAST. Their potency of irreversible inhibition of serine proteases is described in different enzyme assays using chymotrypsin leading to binding affinities up to 22 μM.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride solution, ~0.1 M in ethanol (T)
Pricing and availability is not currently available.