Merck
  • Synthesis of new glycyrrhetinic acid derived ring A azepanone, 29-urea and 29-hydroxamic acid derivatives as selective 11β-hydroxysteroid dehydrogenase 2 inhibitors.
Synthesis of new glycyrrhetinic acid derived ring A azepanone, 29-urea and 29-hydroxamic acid derivatives as selective 11β-hydroxysteroid dehydrogenase 2 inhibitors.
Bioorganic & medicinal chemistry (2011-03-08)
Rawindra Gaware, Rupesh Khunt, Laszlo Czollner, Christian Stanetty, Thierry Da Cunha, Denise V Kratschmar, Alex Odermatt, Paul Kosma, Ulrich Jordis, Dirk Classen-Houben
ABSTRACT
Glycyrrhetinic acid, the metabolite of the natural product glycyrrhizin, is a well known nonselective inhibitor of 11β-hydroxysteroid dehydrogenase (11β-HSD) type 1 and type 2. Whereas inhibition of 11β-HSD1 is currently under consideration for treatment of metabolic diseases, such as obesity and diabetes, 11β-HSD2 inhibitors may find therapeutic applications in chronic inflammatory diseases and certain forms of cancer. Recently, we published a series of hydroxamic acid derivatives of glycyrrhetinic acid showing high selectivity for 11β-HSD2. The most potent and selective compound is active against human 11β-HSD2 in the low nanomolar range with a 350-fold selectivity over human 11β-HSD1. Starting from the lead compounds glycyrrhetinic acid and the hydroxamic acid derivatives, novel triterpene type derivatives were synthesized and analyzed for their biological activity against overexpressed human 11β-HSD1 and 11β-HSD2 in cell lysates. Here we describe novel 29-urea- and 29-hydroxamic acid derivatives of glycyrrhetinic acid as well as derivatives with the Beckman rearrangement of the 3-oxime to a seven-membered ring, and the rearrangement of the C-ring from 11-keto-12-ene to 12-keto-9(11)-ene. The combination of modifications on different positions led to compounds comprising further improved selective inhibition of 11β-HSD2 in the lower nanomolar range with up to 3600-fold selectivity.
Sigma-Aldrich
18β-Glycyrrhetinic acid, 97%
Pricing and availability is not currently available.